Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153391.2 | NP_700440.2 | 34 - 35 | 3904 | 84 | |
GenBank mRNA | AK129413.1 | - | 25 - 26 | 4276 | 84 | |
AK147970.1 | - | 34 - 35 | 3904 | 84 | ||
AY029258.1 | - | 34 - 35 | 3974 | 84 | ||
BC046432.1 | - | 19 - 20 | 2104 | 84 | ||
BC060134.1 | - | 34 - 35 | 3864 | 84 | ||
BC082301.1 | - | 32 - 33 | 5336 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |