Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 14 - 15 | 2435 | 66 | |
NM_001163025.1 | NP_001156497.1 | 14 - 15 | 2438 | 66 | ||
NM_153395.2 | NP_700444.2 | 14 - 15 | 2435 | 66 | ||
GenBank mRNA | AB014059.1 | - | 14 - 15 | 1957 | 66 | |
AK122425.1 | - | 14 - 15 | 2435 | 66 | ||
AK163740.1 | - | 2 - 3 | 2279 | 66 | ||
BC048406.1 | - | 1 - 2 | 211 | 66 | ||
BC150943.1 | - | 14 - 15 | 1959 | 9087854 | 66 | |
CB244438.1 | - | 3 - 4 | 333 | 66 | ||
CN525456.1 | - | 3 - 4 | 386 | 66 | ||
CX122556.1 | - | 1 - 2 | 49 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |