Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153415.3 | NP_700464.2 | 6 - 7 | 1302 | 75 | |
GenBank mRNA | AF246235.1 | - | 6 - 7 | 906 | 75 | |
AK043573.1 | - | 6 - 7 | 1003 | 75 | ||
AK043713.1 | - | 6 - 7 | 844 | 75 | ||
AK050915.1 | - | 6 - 7 | 871 | 75 | ||
AK076813.1 | - | 6 - 7 | 1304 | 75 | ||
AK085703.1 | - | 6 - 7 | 1371 | 75 | ||
AY090481.1 | - | 6 - 7 | 1302 | 75 | ||
AY090482.1 | - | 6 - 7 | 902 | 75 | ||
BC052045.1 | - | 6 - 7 | 874 | 6402328 | 75 | |
BC062965.1 | - | 6 - 7 | 874 | 75 | ||
BC079589.1 | - | 6 - 7 | 872 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |