Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153416.2 | NP_700465.2 | 5 - 6 | 591 | 62 | |
GenBank mRNA | AK034591.1 | - | 5 - 6 | 486 | 62 | |
AK079849.1 | - | 5 - 6 | 528 | 62 | ||
AK083401.1 | - | 5 - 6 | 504 | 62 | ||
AK083537.1 | - | 5 - 6 | 512 | 62 | ||
AK087134.1 | - | 5 - 6 | 567 | 62 | ||
AK088247.1 | - | 5 - 6 | 508 | 62 | ||
AK134406.1 | - | 5 - 6 | 482 | 62 | ||
AK165243.1 | - | 5 - 6 | 506 | 62 | ||
AK166118.1 | - | 5 - 6 | 506 | 62 | ||
AK169523.1 | - | 5 - 6 | 543 | 62 | ||
BC018191.1 | - | 5 - 6 | 492 | 3994248 | 62 | |
BC025501.1 | - | 5 - 6 | 538 | 5321013 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |