Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165954.1 | NP_001159426.1 | 12 - 13 | 2487 | 86 | |
NM_001165955.1 | NP_001159427.1 | 11 - 12 | 2403 | 86 | ||
NM_001165956.1 | NP_001159428.1 | 13 - 14 | 2361 | 86 | ||
NM_153421.2 | NP_700470.2 | 12 - 13 | 2502 | 86 | ||
GenBank mRNA | AJ414610.1 | - | 12 - 13 | 2488 | 86 | |
AK037348.1 | - | 13 - 14 | 2547 | 86 | ||
AK087339.1 | - | 11 - 12 | 2397 | 86 | ||
AK166208.1 | - | 12 - 13 | 2282 | 86 | ||
BC044839.1 | - | 5 - 6 | 1188 | 86 | ||
BC132533.1 | - | 12 - 13 | 2471 | 40130810 | 86 | |
BC145123.1 | - | 13 - 14 | 2345 | 9053671 | 86 | |
BC145124.1 | - | 13 - 14 | 2345 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |