Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gramd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172107.1 | NP_001165578.1 | 5 - 6 | 655 | 93 | |
NM_153528.2 | NP_705756.1 | 5 - 6 | 885 | 93 | ||
GenBank mRNA | AK029543.1 | - | 5 - 6 | 656 | 93 | |
AK035257.1 | - | 5 - 6 | 93 | |||
AK129016.1 | - | 5 - 6 | 824 | 93 | ||
BC037472.1 | - | 5 - 6 | 819 | 4236901 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |