Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153555.2 | NP_705783.1 | 11 - 12 | 1668 | 111 | |
GenBank mRNA | AK035701.1 | - | 11 - 12 | 1567 | 111 | |
AK098114.1 | - | 11 - 12 | 1687 | 111 | ||
AK146825.1 | - | 11 - 12 | 1595 | 111 | ||
BC023804.1 | - | 12 - 13 | 1722 | 5352941 | 111 | |
BC078641.1 | - | 11 - 12 | 2580 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |