Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153558.1 | NP_705786.1 | 1 - 2 | 107 | 66 | |
GenBank mRNA | AY360148.1 | - | 1 - 2 | 143 | 66 | |
BC027556.1 | - | 1 - 2 | 107 | 1434737 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |