Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153805.1 | NP_722500.1 | 14 - 15 | 1698 | 95 | |
GenBank mRNA | AK142182.1 | - | 13 - 14 | 2214 | 95 | |
AK154801.1 | - | 13 - 14 | 2235 | 95 | ||
BC034126.1 | - | 14 - 15 | 1698 | 4912536 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |