Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007496.2 | NP_031522.2 | 9 - 10 | 10184 | 64 | |
GenBank mRNA | AK220555.1 | - | 1 - 2 | 1644 | 64 | |
BC060729.1 | - | 1 - 2 | 3403 | 64 | ||
BC082769.1 | - | 1 - 2 | 1119 | 64 | ||
D26046.1 | - | 9 - 10 | 10181 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |