Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007567.2 | NP_031593.2 | 2 - 3 | 748 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |