Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081001.2 | NP_001074470.1 | 16 - 17 | 8074 | 87 | |
NM_009765.3 | NP_033895.2 | 16 - 17 | 7880 | 87 | ||
GenBank mRNA | AK165522.1 | - | 6 - 7 | 1209 | 87 | |
BC059904.1 | - | 6 - 7 | 1337 | 87 | ||
U65594.1 | - | 16 - 17 | 7857 | 87 | ||
U82270.1 | - | 15 - 16 | 7566 | 87 | ||
U89652.1 | - | 15 - 16 | 7604 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |