Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011082.3 | NP_035212.2 | 3 - 4 | 570 | 71 | |
GenBank mRNA | AK128944.1 | - | 3 - 4 | 570 | 71 | |
AK142656.1 | - | 3 - 4 | 572 | 71 | ||
BC013556.1 | - | 3 - 4 | 503 | 3601095 | 71 | |
U06431.1 | - | 3 - 4 | 474 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |