Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013569.2 | NP_038597.2 | 2 - 3 | 629 | 62 | |
GenBank mRNA | AF012868.1 | - | 2 - 3 | 635 | 62 | |
AY374424.1 | - | 2 - 3 | 314 | 62 | ||
BC051016.1 | - | 2 - 3 | 460 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |