Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2108248 |
AK019467.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2108248 |
BC046306.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2108248 |
NM_026639.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2054854 |
AK004744.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2054854 |
AK019467.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2054854 |
BC046306.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2054854 |
NM_026639.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
428803 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
AK050597.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
AY163161.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
BC117052.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
BC119354.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
M92418.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
NM_001082545.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580655 |
NM_173869.3 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
731322 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
891058 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
806643 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
20687 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
77546 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P141720 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2054854 |
AK004744.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2054854 |
AK019467.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2054854 |
BC046306.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2054854 |
NM_026639.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2108248 |
AK019467.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2108248 |
BC046306.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2108248 |
NM_026639.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P450191 |
AK019467.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P450191 |
BC046306.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P450191 |
NM_026639.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874847 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874837 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874836 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874873 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874862 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874859 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874835 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874868 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874864 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874863 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874866 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874849 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874839 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874852 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874853 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874855 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874858 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874841 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874851 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874870 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874831 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874872 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874838 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874875 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874844 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874828 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874856 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874843 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24874826 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01453444 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P141720 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P450191 |
AK019467.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P450191 |
BC046306.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P450191 |
NM_026639.2 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129612 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P01453444 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P141720 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05815566 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05089588 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05089581 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05089585 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05089583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874852 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874789 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874760 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874851 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874824 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874856 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874742 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874876 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874738 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874814 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874766 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874736 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874872 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874844 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874904 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874849 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874837 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874830 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874732 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874748 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874734 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874822 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874740 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874826 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874847 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874743 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874889 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874772 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874866 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874750 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874819 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874729 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874815 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874864 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874895 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874784 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874791 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24874875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874835 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05574629 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874883 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05060268 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874814 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874852 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05619651 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874817 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874901 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874837 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874897 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874871 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874903 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874872 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874860 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874892 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874866 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874856 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874888 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05641147 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874793 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874828 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874791 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874874 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874870 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874862 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05619650 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874831 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874806 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874826 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874854 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05060270 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874836 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874813 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874904 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874868 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874900 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05619657 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874864 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874819 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874895 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05082944 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05126934 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874876 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874830 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874794 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874815 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874849 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874873 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874851 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874844 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05397407 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874886 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874902 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24874825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05574630 |
- |
- |