Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027141.2 | NP_081417.2 | 1 - 2 | 109 | 64 | |
GenBank mRNA | AF403038.1 | - | 1 - 2 | 109 | 64 | |
AK009325.1 | - | 1 - 2 | 109 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |