Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spsb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163750.1 | NP_001157222.1 | 5 - 6 | 1041 | 101 | |
NM_001163751.1 | NP_001157223.1 | 5 - 6 | 696 | 101 | ||
NM_027141.2 | NP_081417.2 | 5 - 6 | 717 | 101 | ||
GenBank mRNA | AF403038.1 | - | 5 - 6 | 717 | 101 | |
AK009325.1 | - | 5 - 6 | 717 | 101 | ||
AK076170.1 | - | 5 - 6 | 993 | 101 | ||
AK082768.1 | - | 5 - 6 | 640 | 101 | ||
AK167474.1 | - | 4 - 5 | 1034 | 101 | ||
AK220234.1 | - | 5 - 6 | 979 | 101 | ||
BC026523.1 | - | 5 - 6 | 1382 | 101 | ||
BC072556.1 | - | 5 - 6 | 1002 | 5691114 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |