Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286260.1 | NP_001273189.1 | 10 - 11 | 2074 | 136 | |
NM_018823.2 | NP_061293.2 | 11 - 12 | 2128 | 136 | ||
NM_133957.3 | NP_598718.2 | 12 - 13 | 2190 | 136 | ||
GenBank mRNA | AF162853.1 | - | 10 - 11 | 1985 | 136 | |
AF200687.1 | - | 11 - 12 | 1763 | 136 | ||
AF228706.1 | - | 8 - 9 | 1828 | 136 | ||
AF369980.1 | - | 12 - 13 | 1829 | 136 | ||
AF453571.2 | - | 12 - 13 | 1798 | 136 | ||
AK122381.1 | - | 2 - 3 | 215 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |