Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145537.2 | NP_663512.2 | 1 - 2 | 244 | 93 | |
GenBank mRNA | AK029164.1 | - | 1 - 2 | 195 | 93 | |
AK075650.1 | - | 1 - 2 | 158 | 93 | ||
AK079296.1 | - | 1 - 2 | 180 | 93 | ||
AK080636.1 | - | 1 - 2 | 189 | 93 | ||
AK149892.1 | - | 1 - 2 | 180 | 93 | ||
AK155024.1 | - | 1 - 2 | 207 | 93 | ||
AK166998.1 | - | 1 - 2 | 180 | 93 | ||
BC008268.1 | - | 1 - 2 | 155 | 2811074 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |