Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153166.2 | NP_694806.1 | 17 - 18 | 1413 | 74 | |
GenBank mRNA | AK044042.1 | - | 17 - 18 | 1339 | 74 | |
BC036971.1 | - | 17 - 18 | 1424 | 4527212 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |