Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153574.2 | NP_705802.1 | 12 - 13 | 1555 | 118 | |
GenBank mRNA | AJ510136.1 | - | 12 - 13 | 1541 | 118 | |
AK036291.1 | - | 5 - 6 | 762 | 118 | ||
AK052421.1 | - | 12 - 13 | 1496 | 118 | ||
BC005554.1 | - | 12 - 13 | 1460 | 118 | ||
BC054053.1 | - | 12 - 13 | 1529 | 118 | ||
BC060978.1 | - | 12 - 13 | 1492 | 118 | ||
BC080861.1 | - | 12 - 13 | 1555 | 30357292 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |