Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
NM_001252684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
NM_001252684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
NM_001252685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
NM_001252685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
NM_153808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
NM_153808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011444.mm.1 |
TC1900001265.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011446.mm.1 |
NM_001252685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011446.mm.1 |
NM_001252685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011446.mm.1 |
TC1900001265.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
NM_001252684.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
NM_001252684.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
NM_001252685.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
NM_001252685.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
NM_153808.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
NM_153808.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900011443.mm.1 |
TC1900001265.mm.1 |
MTA TC |