Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mycbpap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170671.2 | NP_733771.2 | 5 - 6 | 681 | 60 | |
GenBank mRNA | BC027327.1 | - | 1 - 2 | 42 | 3991131 | 60 |
BU840494.1 | - | 5 - 6 | 681 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |