Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
NM_001146348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
NM_001146348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
NM_001146350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
NM_001146350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
NM_007932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
NM_007932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005392.mm.1 |
TC0200005472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
NM_001146348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
NM_001146348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
NM_001146350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
NM_001146350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
NM_007932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
NM_007932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005395.mm.1 |
TC0200005472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
NM_001146348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
NM_001146348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
NM_001146350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
NM_001146350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
NM_007932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
NM_007932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046657.mm.1 |
TC0200005472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
NM_001146348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
NM_001146348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
NM_001146350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
NM_001146350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
NM_007932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
NM_007932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005391.mm.1 |
TC0200005472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
NM_001146348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
NM_001146348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
NM_001146350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
NM_001146350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
NM_007932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
NM_007932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046658.mm.1 |
TC0200005472.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
NM_001146348.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
NM_001146348.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
NM_001146350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
NM_001146350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
NM_007932.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
NM_007932.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005396.mm.1 |
TC0200005472.mm.1 |
MTA TC |