Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146348.1 | NP_001139820.1 | 9 - 10 | 1632 | 69 | |
NM_001146350.1 | NP_001139822.1 | 9 - 10 | 1635 | 69 | ||
NM_007932.2 | NP_031958.2 | 9 - 10 | 1635 | 69 | ||
GenBank mRNA | AK151302.1 | - | 9 - 10 | 1605 | 69 | |
AY679531.1 | - | 9 - 10 | 1635 | 69 | ||
BC029080.1 | - | 9 - 10 | 1549 | 4168250 | 69 | |
S69407.1 | - | 10 - 11 | 1500 | 69 | ||
X77952.1 | - | 9 - 10 | 1635 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |