Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007934.3 | NP_031960.1 | 5 - 6 | 1438 | 77 | |
GenBank mRNA | AK031439.1 | - | 4 - 5 | 470 | 77 | |
AK034613.1 | - | 5 - 6 | 1410 | 77 | ||
AK128975.1 | - | 5 - 6 | 1395 | 77 | ||
AK146808.1 | - | 5 - 6 | 1410 | 77 | ||
BC127060.1 | - | 5 - 6 | 1302 | 40109863 | 77 | |
DQ000292.1 | - | 5 - 6 | 1178 | 77 | ||
M29961.1 | - | 5 - 6 | 1257 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |