Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963727 |
NM_007947.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963727 |
AK136629.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963727 |
AK162355.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963727 |
U68546.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1963727 |
NM_001042630.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
AF151691.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
AK006905.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
AK145244.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
AK154537.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
AY515316.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
AY515317.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
BC057575.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
BC082278.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
837665 |
NM_011972.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
564555 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P128366 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963727 |
NM_007947.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963727 |
AK136629.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963727 |
AK162355.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963727 |
U68546.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1963727 |
NM_001042630.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P191875 |
NM_007947.2 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P191875 |
U68546.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073643 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073624 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073649 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073623 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073645 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073651 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073635 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073638 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073625 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073616 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073627 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073647 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073634 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073640 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073617 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073619 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073622 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073648 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073644 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073629 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073621 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073641 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073637 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073630 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21073633 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P128366 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P191875 |
NM_007947.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P191875 |
U68546.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P112909 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121716 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P128366 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01486572 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01486568 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01486596 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01486566 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01486576 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01486578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073648 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073579 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073674 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073644 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073559 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073600 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073669 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073655 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073625 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073659 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073582 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073567 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073589 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073671 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073587 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073597 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073677 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073662 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073676 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073558 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073658 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073627 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073616 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073570 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073576 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073660 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073678 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073635 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073675 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073574 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073651 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073645 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073640 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073555 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21073562 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073624 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073627 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01486578 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02128529 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02186804 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073677 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073645 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073643 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073682 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02186806 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073635 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073651 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073641 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01522237 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073629 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073653 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073656 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073616 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01486567 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073622 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073661 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073633 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01522240 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01872577 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02186808 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01522227 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01522248 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073659 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073634 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073666 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073621 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073680 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073658 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073676 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073630 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01522250 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073673 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073647 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073675 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073660 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073619 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073650 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073674 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01486587 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21073678 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01486592 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02186803 |
- |
- |