Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 14 - 15 | 3063 | 58 | |
GenBank mRNA | AF300701.1 | - | 13 - 14 | 2540 | 58 | |
AK156112.1 | - | 14 - 15 | 3065 | 58 | ||
U36488.1 | - | 10 - 11 | 2225 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons |