Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007955.3 | NP_031981.3 | 16 - 17 | 3620 | 106 | |
GenBank mRNA | AF300701.1 | - | 15 - 16 | 3097 | 106 | |
AK156112.1 | - | 16 - 17 | 3623 | 106 | ||
U36488.1 | - | 12 - 13 | 2782 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons |