Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00019114.mm.1 |
NM_007957.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019114.mm.1 |
NM_007957.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019114.mm.1 |
TC0X00003046.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019113.mm.1 |
NM_007957.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019113.mm.1 |
NM_007957.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019113.mm.1 |
TC0X00003046.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019116.mm.1 |
NM_007957.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019116.mm.1 |
NM_007957.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019116.mm.1 |
TC0X00003046.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019117.mm.1 |
NM_007957.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019117.mm.1 |
NM_007957.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019117.mm.1 |
TC0X00003046.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019112.mm.1 |
NM_007957.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019112.mm.1 |
NM_007957.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019112.mm.1 |
TC0X00003046.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019111.mm.1 |
NM_007957.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019111.mm.1 |
NM_007957.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019111.mm.1 |
TC0X00003046.mm.1 |
MTA TC |