Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_001163394.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_001163394.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_001163395.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_001163395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_001163396.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_001163396.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_007965.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
NM_007965.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007920.mm.1 |
TC1200001110.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007917.mm.1 |
NM_001163396.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007917.mm.1 |
NM_001163396.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007917.mm.1 |
TC1200001110.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007914.mm.1 |
NM_001163394.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007914.mm.1 |
NM_001163394.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007914.mm.1 |
NM_007965.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007914.mm.1 |
NM_007965.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007914.mm.1 |
TC1200001110.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200007916.mm.1 |
NM_001163396.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007916.mm.1 |
NM_001163396.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200007916.mm.1 |
TC1200001110.mm.1 |
MTA TC |