Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146689.1 | NP_001140161.1 | 4 - 5 | 477 | 97 | |
NM_007971.2 | NP_031997.2 | 4 - 5 | 504 | 97 | ||
GenBank mRNA | AK088575.1 | - | 4 - 5 | 498 | 97 | |
AK153835.1 | - | 3 - 4 | 425 | 97 | ||
AK157853.1 | - | 4 - 5 | 505 | 97 | ||
AK220174.1 | - | 4 - 5 | 459 | 97 | ||
BC079538.1 | - | 4 - 5 | 428 | 6817366 | 97 | |
U52951.1 | - | 4 - 5 | 494 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |