Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146689.1 | NP_001140161.1 | 15 - 16 | 1973 | 104 | |
NM_007971.2 | NP_031997.2 | 15 - 16 | 1985 | 104 | ||
GenBank mRNA | AK157853.1 | - | 15 - 16 | 1986 | 104 | |
AK220174.1 | - | 15 - 16 | 1940 | 104 | ||
BC003772.1 | - | 15 - 16 | 1894 | 3492110 | 104 | |
BC016391.1 | - | 15 - 16 | 1907 | 3586689 | 104 | |
BC079538.1 | - | 15 - 16 | 1924 | 6817366 | 104 | |
U52951.1 | - | 15 - 16 | 1975 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |