Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143616.1 | - | 7 - 8 | 957 | 109 | |
AK144669.1 | - | 7 - 8 | 964 | 109 | ||
AK146813.1 | - | 7 - 8 | 926 | 109 | ||
AK146918.1 | - | 7 - 8 | 934 | 109 | ||
BC003741.1 | - | 7 - 8 | 923 | 3488803 | 109 | |
D63850.1 | - | 7 - 8 | 940 | 109 | ||
FN687734.2 | - | 7 - 8 | 683 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |