Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hipk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145824.1 | NP_001139296.1 | 8 - 9 | 2432 | 120 | |
NM_010434.2 | NP_034564.2 | 8 - 9 | 2432 | 120 | ||
GenBank mRNA | AF077660.1 | - | 8 - 9 | 2186 | 120 | |
AF170305.1 | - | 7 - 8 | 1896 | 120 | ||
AF305238.1 | - | 7 - 8 | 1894 | 120 | ||
AK140130.1 | - | 8 - 9 | 2535 | 120 | ||
BC131663.1 | - | 8 - 9 | 2064 | 120 | ||
BC131664.1 | - | 8 - 9 | 2063 | 40110751 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |