Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmg20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163165.1 | NP_001156637.1 | 2 - 3 | 212 | 79 | |
NM_001163166.1 | NP_001156638.1 | 2 - 3 | 109 | 79 | ||
NM_010440.3 | NP_034570.1 | 1 - 2 | 296 | 79 | ||
GenBank mRNA | AF146224.1 | - | 1 - 2 | 279 | 79 | |
AK153816.1 | - | 2 - 3 | 110 | 79 | ||
AK155615.1 | - | 2 - 3 | 213 | 79 | ||
AK155652.1 | - | 2 - 3 | 114 | 79 | ||
AK155890.1 | - | 1 - 2 | 172 | 79 | ||
AL355734.1 | - | 1 - 2 | 291 | 79 | ||
BC011334.1 | - | 2 - 3 | 145 | 4012515 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |