Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013547.3 | NP_038575.2 | 3 - 4 | 345 | 85 | |
GenBank mRNA | AK002584.1 | - | 2 - 3 | 217 | 85 | |
AK050193.1 | - | 3 - 4 | 245 | 85 | ||
BC037628.1 | - | 3 - 4 | 259 | 4973560 | 85 | |
BC055029.1 | - | 3 - 4 | 226 | 5054479 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |