Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015774.3 | NP_056589.1 | 13 - 14 | 1195 | 116 | |
GenBank mRNA | AK009667.1 | - | 3 - 4 | 420 | 116 | |
AK142595.1 | - | 13 - 14 | 1186 | 116 | ||
BC025102.1 | - | 13 - 14 | 1147 | 2654098 | 116 | |
CT010195.1 | - | 13 - 14 | 1113 | 116 | ||
CT010259.1 | - | 13 - 14 | 1113 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |