Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700021530.mm.1 |
TC0700002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021530.mm.1 |
NM_001162950.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021530.mm.1 |
NM_001162950.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021530.mm.1 |
NM_016868.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021530.mm.1 |
NM_016868.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021533.mm.1 |
TC0700002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021533.mm.1 |
NM_001162950.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021533.mm.1 |
NM_001162950.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021533.mm.1 |
NM_016868.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021533.mm.1 |
NM_016868.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021534.mm.1 |
TC0700002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021534.mm.1 |
NM_001162950.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021534.mm.1 |
NM_001162950.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021534.mm.1 |
NM_016868.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021534.mm.1 |
NM_016868.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021532.mm.1 |
TC0700002445.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700021532.mm.1 |
NM_001162950.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021532.mm.1 |
NM_001162950.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021532.mm.1 |
NM_016868.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700021532.mm.1 |
NM_016868.3 |
Refseq |