Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021398.3 | NP_067373.2 | 5 - 6 | 808 | 72 | |
GenBank mRNA | AF188622.1 | - | 5 - 6 | 802 | 72 | |
AK086962.1 | - | 4 - 5 | 1237 | 72 | ||
BC089603.1 | - | 4 - 5 | 508 | 6758493 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |