Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myoz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021508.3 | NP_067483.1 | 1 - 2 | 115 | 91 | |
GenBank mRNA | AJ005620.1 | - | 1 - 2 | 55 | 91 | |
AK009077.1 | - | 1 - 2 | 79 | 91 | ||
AK041896.1 | - | 1 - 2 | 71 | 91 | ||
AY013298.1 | - | 1 - 2 | 33 | 91 | ||
BC024660.1 | - | 1 - 2 | 171 | 3967841 | 91 | |
BY790048.1 | - | 1 - 2 | 116 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |