Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinagl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168333.1 | NP_001161805.1 | 5 - 6 | 669 | 80 | |
NM_023476.3 | NP_075965.2 | 5 - 6 | 810 | 80 | ||
GenBank mRNA | AB047402.1 | - | 5 - 6 | 811 | 80 | |
AB050626.1 | - | 5 - 6 | 647 | 80 | ||
AK087142.1 | - | 81 | ||||
BC005738.1 | - | 5 - 6 | 637 | 2650851 | 80 | |
BC018539.1 | - | 5 - 6 | 654 | 3596560 | 80 | |
CT010275.1 | - | 4 - 5 | 428 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |