Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290521.1 | NP_001277450.1 | 4 - 5 | 558 | 89 | |
NM_001290522.1 | NP_001277451.1 | 4 - 5 | 448 | 89 | ||
NM_023597.2 | NP_076086.1 | 4 - 5 | 451 | 89 | ||
GenBank mRNA | AF274321.1 | - | 4 - 5 | 428 | 89 | |
AK017259.1 | - | 4 - 5 | 441 | 89 | ||
AK141026.1 | - | 4 - 5 | 556 | 89 | ||
BC026500.1 | - | 3 - 4 | 337 | 89 | ||
BC037102.1 | - | 4 - 5 | 441 | 5364307 | 89 | |
CK618879.1 | - | 4 - 5 | 558 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |