Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964348 |
AK086508.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964348 |
BC023365.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964348 |
BC038274.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964348 |
AK018634.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1964348 |
NM_023608.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
85458 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921232 |
AF213381.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921232 |
AK003527.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921232 |
AK030626.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921232 |
BC032923.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
921232 |
NM_013852.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
490428 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
851985 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
542833 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
368025 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
709986 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
89519 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P107033 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964348 |
AK086508.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964348 |
BC023365.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964348 |
BC038274.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964348 |
AK018634.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1964348 |
NM_023608.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355957 |
AK086508.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355957 |
BC038274.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355957 |
AK018634.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355957 |
NM_023608.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P355957 |
AB048364.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561065 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561034 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561045 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561053 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561036 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561056 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561061 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561031 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561066 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561028 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561062 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561041 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561054 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561030 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561051 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561042 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561044 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561049 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561038 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561032 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561064 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561060 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561057 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561040 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561050 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P32561047 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09374309 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P107033 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P355957 |
AK086508.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P355957 |
BC038274.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P355957 |
AK018634.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P355957 |
NM_023608.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P355957 |
AB048364.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P122761 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P107033 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032665 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032682 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032684 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032683 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032668 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032670 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P15032664 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561080 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561105 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561040 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561138 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561053 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561113 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561145 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561115 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561143 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561047 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561127 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561049 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561060 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561050 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561086 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561110 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P32561102 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561038 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15032676 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561097 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15096695 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15052190 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561049 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561123 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561061 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14979803 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14725213 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561034 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561066 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15153804 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561042 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561082 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561051 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14878214 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14979779 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561091 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561060 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561086 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15052189 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561149 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14823767 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561140 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15096693 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561117 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561152 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561083 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561138 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15012188 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15125776 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561032 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15032684 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561057 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14600478 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561146 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561103 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561132 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561041 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14979771 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14979772 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561092 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561134 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561047 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561148 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561093 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15125772 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561081 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561145 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561064 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561119 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561107 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15125770 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561045 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561121 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561062 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561113 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561143 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15012186 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14823765 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561073 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14979773 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14979778 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561076 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561040 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561078 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561108 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561084 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561106 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561104 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P32561036 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P14878220 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P15032689 |
- |
- |