Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023732.2 | NP_076221.1 | 4 - 5 | 1135 | 78 | |
GenBank mRNA | AF045562.1 | - | 4 - 5 | 997 | 78 | |
AK004605.1 | - | 4 - 5 | 1215 | 78 | ||
AK040323.1 | - | 4 - 5 | 1139 | 78 | ||
AK151165.1 | - | 4 - 5 | 1244 | 78 | ||
AK164003.1 | - | 4 - 5 | 1139 | 78 | ||
AK168642.1 | - | 4 - 5 | 1236 | 78 | ||
BC006634.1 | - | 4 - 5 | 1135 | 3710552 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |