Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023733.3 | NP_076222.1 | 5 - 6 | 575 | 115 | |
GenBank mRNA | AK004567.2 | - | 5 - 6 | 576 | 115 | |
AK037973.1 | - | 4 - 5 | 1027 | 115 | ||
BC006593.1 | - | 5 - 6 | 563 | 3156501 | 115 | |
BC012308.1 | - | 5 - 6 | 590 | 3500167 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |