Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023773.2 | NP_076262.2 | 8 - 9 | 2027 | 74 | |
GenBank mRNA | AB041598.2 | - | 8 - 9 | 2028 | 74 | |
AK016067.1 | - | 3 - 4 | 272 | 74 | ||
AK148654.1 | - | 1 - 2 | 30 | 74 | ||
AK158773.1 | - | 8 - 9 | 2005 | 74 | ||
AK159495.1 | - | 8 - 9 | 2005 | 74 | ||
BC079558.1 | - | 8 - 9 | 1959 | 74 | ||
BC145164.1 | - | 8 - 9 | 1952 | 74 | ||
BC145700.1 | - | 8 - 9 | 1952 | 40131003 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |