Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023844.5 | NP_076333.3 | 3 - 4 | 607 | 143 | |
GenBank mRNA | AF255911.1 | - | 3 - 4 | 330 | 143 | |
AJ291757.1 | - | 3 - 4 | 543 | 143 | ||
AK010616.1 | - | 3 - 4 | 449 | 143 | ||
AK010826.1 | - | 3 - 4 | 451 | 143 | ||
AK013914.1 | - | 3 - 4 | 448 | 143 | ||
AK028757.1 | - | 3 - 4 | 298 | 143 | ||
AK045095.1 | - | 3 - 4 | 345 | 143 | ||
AK078128.1 | - | 3 - 4 | 333 | 143 | ||
AK153636.1 | - | 3 - 4 | 297 | 143 | ||
BC028778.1 | - | 3 - 4 | 607 | 1195543 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |