Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170489.1 | NP_001163960.1 | 7 - 8 | 1359 | 70 | |
NM_024251.4 | NP_077213.2 | 7 - 8 | 1229 | 70 | ||
GenBank mRNA | AK008513.1 | - | 7 - 8 | 1360 | 70 | |
AK034191.1 | - | 7 - 8 | 1209 | 70 | ||
BC002179.1 | - | 7 - 8 | 1007 | 3487226 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |