Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mansc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026345.4 | NP_080621.1 | 2 - 3 | 592 | 70 | |
GenBank mRNA | AK002644.1 | - | 2 - 3 | 501 | 70 | |
AK018635.1 | - | 2 - 3 | 483 | 70 | ||
AK018660.1 | - | 2 - 3 | 505 | 70 | ||
AK033526.1 | - | 2 - 3 | 504 | 70 | ||
AK033557.1 | - | 2 - 3 | 593 | 70 | ||
AK046837.1 | - | 2 - 3 | 481 | 70 | ||
AK171842.1 | - | 2 - 3 | 509 | 70 | ||
BC039930.1 | - | 2 - 3 | 463 | 3598325 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |